siRNA Details
| Gene Symbol: Azin2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC078981.1 | - | 349 | 7112100 | ||
| CK470797.1 | - | 553 | ||||
| HQ413774.1 | - | 2 | 198 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Azin2| Gene Symbol: Azin2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC078981.1 | - | 349 | 7112100 | ||
| CK470797.1 | - | 553 | ||||
| HQ413774.1 | - | 2 | 198 | |||