siRNA Details
| Gene Symbol: Efcab11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024350.1 | NP_001019521.1 | 4, 5 | 388 | ||
| XM_006240446.2 | XP_006240508.2 | 4, 5 | 490 | |||
| GenBank mRNA | BC079408.1 | - | 4, 5 | 388 | 7113005 | |

Efcab11
Genomic Map