siRNA Details
| Gene Symbol: Nop2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191785.1 | NP_001178714.1 | 16 | 2033 | ||
| Gene Symbol: LOC500881 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008765532.1 | XP_008763754.1 | 2 | 2165 | ||

Nop2
Genomic Map