siRNA Details
| Gene Symbol: Vwc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109312.1 | NP_001102782.1 | 2 | 1087 | ||
| XM_008770349.1 | XP_008768571.1 | 2 | 1374 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vwc2| Gene Symbol: Vwc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109312.1 | NP_001102782.1 | 2 | 1087 | ||
| XM_008770349.1 | XP_008768571.1 | 2 | 1374 | |||