siRNA Details
| Gene Symbol: Zmat5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006251427.2 | XP_006251489.1 | 5 | 1679 | ||
| XM_577351.5 | XP_577351.3 | 5 | 670 | |||
| GenBank mRNA | FQ227485.1 | - | 5 | 422 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zmat5| Gene Symbol: Zmat5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006251427.2 | XP_006251489.1 | 5 | 1679 | ||
| XM_577351.5 | XP_577351.3 | 5 | 670 | |||
| GenBank mRNA | FQ227485.1 | - | 5 | 422 | ||