siRNA Details
| Gene Symbol: LOC502176 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025057.2 | NP_001020228.2 | 2 | 165 | ||
| GenBank mRNA | AY325157.1 | - | 2 | 165 | ||
| Gene Symbol: Acat2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AY325187.1 | - | 1913 | |||
| Gene Symbol: RGD1563400 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008769080.1 | XP_008767302.1 | 1 | 154 | ||
| Gene Symbol: Tmem170b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AY310151.1 | - | 3 | 210 | ||

LOC502176
Genomic Map