siRNA Details
| Gene Symbol: Tmem181 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130939.1 | NP_001124411.1 | 6 | 526 | ||
| GenBank mRNA | BC168231.1 | - | 6 | 526 | 7387910 | |
| FQ211485.1 | - | 78 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem181| Gene Symbol: Tmem181 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130939.1 | NP_001124411.1 | 6 | 526 | ||
| GenBank mRNA | BC168231.1 | - | 6 | 526 | 7387910 | |
| FQ211485.1 | - | 78 | ||||