siRNA Details
| Gene Symbol: H1foo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109351.1 | NP_001102821.1 | 3 | 537 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

H1foo| Gene Symbol: H1foo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109351.1 | NP_001102821.1 | 3 | 537 | ||