siRNA Details
| Gene Symbol: Clec7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001173386.1 | NP_001166857.1 | 5 | 569 | ||
| XM_006237100.2 | XP_006237162.1 | 4 | 557 | |||
| GenBank mRNA | AB455106.1 | - | 5 | 661 | ||
| AB455107.1 | - | 4 | 526 | |||
| BC166774.1 | - | 5 | 569 | |||
| GU357485.1 | - | 5 | 501 | |||

Clec7a
Genomic Map