siRNA Details
| Gene Symbol: Map4k5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001192016.1 | NP_001178945.1 | 24 | 1849 | ||
| XM_006240179.2 | XP_006240241.1 | 25 | 2162 | |||
| XM_006240180.2 | XP_006240242.1 | 24 | 2163 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Map4k5| Gene Symbol: Map4k5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001192016.1 | NP_001178945.1 | 24 | 1849 | ||
| XM_006240179.2 | XP_006240241.1 | 25 | 2162 | |||
| XM_006240180.2 | XP_006240242.1 | 24 | 2163 | |||