siRNA Details
Gene Symbol: Vom2r57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099474.1 | NP_001092944.1 | 4, 5 | 1545 | ||
XM_008765199.1 | XP_008763421.1 | 5, 6 | 1554 |
Gene Symbol: LOC103690080 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008765207.1 | XP_008763429.1 | 4, 5 | 707 |
Gene Symbol: Vom2r56 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099484.1 | NP_001092954.1 | 1545 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
