siRNA Details
| Gene Symbol: Hpcal1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017356.2 | NP_059052.1 | 5 | 865 | ||
| XM_006239914.2 | XP_006239976.1 | 6 | 1472 | |||
| XM_006239916.2 | XP_006239978.1 | 5 | 1367 | |||
| XM_008764605.1 | XP_008762827.1 | 5 | 982 | |||
| XM_008764606.1 | XP_008762828.1 | 7 | 1582 | |||
| GenBank mRNA | BC088759.1 | - | 5 | 865 | 7317905 | |
| D13126.1 | - | 5 | 820 | |||
| FQ214068.1 | - | 3 | 388 | |||

Hpcal1
Genomic Map