siRNA Details
| Gene Symbol: Rab10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017359.2 | NP_059055.2 | 3 | 332 | ||
| GenBank mRNA | AY310140.1 | - | 1 | 96 | ||
| AY724493.1 | - | 3 | 511 | |||
| BC085744.1 | - | 3 | 332 | 7191921 | ||
| M83677.1 | - | 3 | 484 | |||

Rab10
Genomic Map