siRNA Details
| Gene Symbol: Maf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019318.1 | NP_062191.1 | 1 | 1682 | ||
| GenBank mRNA | U56242.1 | - | 1 | 1682 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Maf| Gene Symbol: Maf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019318.1 | NP_062191.1 | 1 | 1682 | ||
| GenBank mRNA | U56242.1 | - | 1 | 1682 | ||