siRNA Details
| Gene Symbol: Plvap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020086.1 | NP_064471.1 | 1 | 271 | ||
| GenBank mRNA | AF154831.1 | - | 1 | 271 | ||
| BC070900.1 | - | 1 | 271 | 7100403 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Plvap| Gene Symbol: Plvap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020086.1 | NP_064471.1 | 1 | 271 | ||
| GenBank mRNA | AF154831.1 | - | 1 | 271 | ||
| BC070900.1 | - | 1 | 271 | 7100403 | ||