siRNA Details
| Gene Symbol: Ndufaf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033971.1 | NP_001029143.1 | 2 | 819 | ||
| NM_020080.2 | NP_064465.1 | 2 | 837 | |||
| XM_006243845.2 | XP_006243907.1 | 3 | 2319 | |||
| GenBank mRNA | BC079306.1 | - | 2 | 571 | 7134644 | |
| FQ216095.1 | - | 2 | 188 | |||
| FQ223486.1 | - | 2 | 132 | |||
| FQ233128.1 | - | 1 | 264 | |||
| U95160.1 | - | 2 | 173 | |||
| U95161.1 | - | 2 | 173 | |||
| U95162.1 | - | 2 | 173 | |||

Ndufaf3
Genomic Map