siRNA Details
| Gene Symbol: Nrbf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022186.1 | NP_071522.1 | 3, 4 | 273 | ||
| XM_008772913.1 | XP_008771135.1 | 2, 3 | 249 | |||
| GenBank mRNA | AB024930.1 | - | 3, 4 | 273 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nrbf2| Gene Symbol: Nrbf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022186.1 | NP_071522.1 | 3, 4 | 273 | ||
| XM_008772913.1 | XP_008771135.1 | 2, 3 | 249 | |||
| GenBank mRNA | AB024930.1 | - | 3, 4 | 273 | ||