siRNA Details
| Gene Symbol: Fxyd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031648.2 | NP_113836.2 | 4, 5 | 265 | ||
| XM_006228838.2 | XP_006228900.1 | 5, 6 | 485 | |||
| XM_006228839.2 | XP_006228901.1 | 4, 5 | 260 | |||
| XM_008759184.1 | XP_008757406.1 | 4, 5 | 274 | |||
| GenBank mRNA | CB712570.1 | - | 4, 5 | 265 | ||
| FM109156.1 | - | 4, 5 | 219 | |||
| FQ211349.1 | - | 4, 5 | 276 | |||
| FQ224131.1 | - | 4, 5 | 228 | |||
| FQ224338.1 | - | 4, 5 | 311 | |||
| U72246.1 | - | 3, 4 | 179 | |||

Fxyd1
Genomic Map