siRNA Details
| Gene Symbol: Rbck1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021764.1 | NP_068532.2 | 3, 4 | 557 | ||
| XM_006235252.2 | XP_006235314.1 | 4, 5 | 482 | |||
| GenBank mRNA | AB011369.1 | - | 3, 4 | 557 | ||
| BC061723.1 | - | 3, 4 | 491 | 5598480 | ||
| U48248.1 | - | 3, 4 | 557 | |||

Rbck1
Genomic Map