siRNA Details
| Gene Symbol: NXPH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007226.2 | NP_009157.1 | 2 | 400 | ||
| GenBank mRNA | AB162675.1 | - | 2 | 400 | ||
| AF043467.1 | - | 2 | 394 | |||
| BC101461.1 | - | 2 | 400 | 40022686 | ||
| BC101462.1 | - | 2 | 400 | 40022687 | ||
| BC101463.1 | - | 2 | 400 | 40022688 | ||
| BC104741.1 | - | 2 | 400 | 40022689 | ||

NXPH2
Genomic Map