siRNA Details
| Gene Symbol: Xiap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022231.2 | NP_071567.1 | 8 | 1697 | ||
| XM_006257506.2 | XP_006257568.1 | 7 | 1523 | |||
| XM_006257508.2 | XP_006257570.1 | 7 | 1637 | |||
| XM_006257509.2 | XP_006257571.1 | 7 | 1538 | |||
| XM_008773529.1 | XP_008771751.1 | 8 | 1664 | |||
| GenBank mRNA | AB033366.1 | - | 8 | 1696 | ||
| AF183429.1 | - | 6 | 1367 | |||
| AF304334.1 | - | 2292 | ||||

Xiap
Genomic Map