siRNA Details
| Gene Symbol: Casp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022260.3 | NP_071596.1 | 4 | 521 | ||
| XM_008760546.1 | XP_008758768.1 | 5 | 1114 | |||
| XM_008760547.1 | XP_008758769.1 | 4 | 1496 | |||
| XM_008760548.1 | XP_008758770.1 | 4 | 823 | |||
| XM_008760549.1 | XP_008758771.1 | 5 | 703 | |||
| XM_008760550.1 | XP_008758772.1 | 4 | 631 | |||
| GenBank mRNA | AF072124.1 | - | 4 | 522 | ||
| BC070936.1 | - | 4 | 522 | 7099111 | ||

Casp7
Genomic Map