siRNA Details
| Gene Symbol: Esm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022604.2 | NP_072126.1 | 1 | 406 | ||
| GenBank mRNA | BC070888.1 | - | 1 | 406 | 7098029 | |
| U80818.1 | - | 1 | 319 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Esm1| Gene Symbol: Esm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022604.2 | NP_072126.1 | 1 | 406 | ||
| GenBank mRNA | BC070888.1 | - | 1 | 406 | 7098029 | |
| U80818.1 | - | 1 | 319 | |||