siRNA Details
| Gene Symbol: Ilkap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022606.1 | NP_072128.1 | 9 | 788 | ||
| XM_008767296.1 | XP_008765518.1 | 8 | 2744 | |||
| XM_008767297.1 | XP_008765519.1 | 7 | 3069 | |||
| GenBank mRNA | AF095927.1 | - | 9 | 788 | ||
| BC062010.1 | - | 9 | 790 | 5621497 | ||

Ilkap
Genomic Map