siRNA Details
| Gene Symbol: Prkab2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022627.2 | NP_072149.1 | 5 | 570 | ||
| XM_006233011.2 | XP_006233073.1 | 6 | 771 | |||
| XM_006233012.2 | XP_006233074.1 | 6 | 671 | |||
| GenBank mRNA | AF182717.1 | - | 5 | 572 | ||

Prkab2
Genomic Map