siRNA Details
| Gene Symbol: Bad | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022698.1 | NP_073189.1 | 3, 4 | 501 | ||
| XM_006230898.2 | XP_006230960.1 | 3, 4 | 399 | |||
| GenBank mRNA | AF003523.1 | - | 3, 4 | 501 | ||
| AF031227.1 | - | 3, 4 | 539 | |||
| AF279910.1 | - | 3, 4 | 501 | |||

Bad
Genomic Map