siRNA Details
| Gene Symbol: Kcnip3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032462.2 | NP_115851.1 | 4 | 348 | ||
| XM_006234989.2 | XP_006235051.1 | 4 | 477 | |||
| XM_006234990.2 | XP_006235052.1 | 3 | 3793 | |||
| GenBank mRNA | AF297118.1 | - | 4 | 350 | ||

Kcnip3
Genomic Map