siRNA Details
| Gene Symbol: Ido1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023973.1 | NP_076463.1 | 10 | 888 | ||
| GenBank mRNA | AF312699.1 | - | 10 | 888 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ido1| Gene Symbol: Ido1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023973.1 | NP_076463.1 | 10 | 888 | ||
| GenBank mRNA | AF312699.1 | - | 10 | 888 | ||