siRNA Details
| Gene Symbol: Chac2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025016.1 | NP_001020187.1 | 3 | 1091 | ||
| GenBank mRNA | BC082031.1 | - | 3 | 1091 | 7130142 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Chac2| Gene Symbol: Chac2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025016.1 | NP_001020187.1 | 3 | 1091 | ||
| GenBank mRNA | BC082031.1 | - | 3 | 1091 | 7130142 | |