siRNA Details
| Gene Symbol: Nol4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107401.1 | NP_001100871.1 | 1 | 461 | ||
| XM_006254453.2 | XP_006254515.1 | 1 | 451 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nol4| Gene Symbol: Nol4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107401.1 | NP_001100871.1 | 1 | 461 | ||
| XM_006254453.2 | XP_006254515.1 | 1 | 451 | |||