siRNA Details
| Gene Symbol: TMEM258 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014206.3 | NP_055021.1 | 3 | 197 | ||
| GenBank mRNA | AF070661.1 | - | 3 | 197 | ||
| AF085986.1 | - | 3 | 170 | |||
| AF086763.1 | - | 3 | 193 | |||
| AK291683.1 | - | 3 | 194 | |||
| BC002750.2 | - | 3 | 168 | 3633279 | ||
| BC015968.1 | - | 3 | 198 | 4073907 | ||

TMEM258
Genomic Map