siRNA Details
Gene Symbol: Cenpw | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001246319.1 | NP_001233248.1 | 2 | 319 | ||
GenBank mRNA | BC129114.1 | - | 2 | 319 | 7112235 | |
FQ220188.1 | - | 1 | 48 | |||
FQ227404.1 | - | 1 | 48 |
Gene Symbol: LOC681382 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_001061496.2 | XP_001061496.1 | 1 | 218 |
