siRNA Details
| Gene Symbol: Jazf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001065610.5 | XP_001065610.1 | 5 | 1192 | ||
| XM_006236535.2 | XP_006236597.1 | 5 | 1980 | |||
| XM_006236536.2 | XP_006236598.1 | 5 | 3098 | |||
| XM_008762953.1 | XP_008761175.1 | 5 | 1319 | |||
| XM_008762954.1 | XP_008761176.1 | 6 | 1534 | |||

Jazf1
Genomic Map