siRNA Details
| Gene Symbol: LOC100360557 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749003.2 | XP_003749051.1 | 1 | 521 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100360557| Gene Symbol: LOC100360557 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749003.2 | XP_003749051.1 | 1 | 521 | ||