siRNA Details
| Gene Symbol: Nudt13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127636.1 | NP_001121108.1 | 4 | 339 | ||
| XM_008770571.1 | XP_008768793.1 | 3 | 286 | |||
| GenBank mRNA | BC166528.1 | - | 4 | 339 | 7189304 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nudt13| Gene Symbol: Nudt13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127636.1 | NP_001121108.1 | 4 | 339 | ||
| XM_008770571.1 | XP_008768793.1 | 3 | 286 | |||
| GenBank mRNA | BC166528.1 | - | 4 | 339 | 7189304 | |