siRNA Details
| Gene Symbol: Fam162a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006248413.2 | XP_006248475.1 | 5 | 608 | ||
| GenBank mRNA | BC097971.1 | - | 5 | 554 | 7442058 | |
| FQ222091.1 | - | 5 | 578 | |||
| M17412.1 | - | 4 | 485 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam162a| Gene Symbol: Fam162a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006248413.2 | XP_006248475.1 | 5 | 608 | ||
| GenBank mRNA | BC097971.1 | - | 5 | 554 | 7442058 | |
| FQ222091.1 | - | 5 | 578 | |||
| M17412.1 | - | 4 | 485 | |||