siRNA Details
| Gene Symbol: XPOT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007235.4 | NP_009166.2 | 6 | 803 | ||
| XM_017018748.1 | XP_016874237.1 | 6 | 663 | |||
| GenBank mRNA | AF039022.1 | - | 5 | 297 | ||
| AK292884.1 | - | 6 | 616 | |||
| AK308108.1 | - | 5 | 554 | |||
| BC020569.1 | - | 5 | 382 | 4420052 | ||
| BQ215290.1 | - | 6 | 723 | |||
| Y16414.1 | - | 5 | 355 | |||

XPOT
Genomic Map