siRNA Details
| Gene Symbol: Fam120a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191816.1 | NP_001178745.1 | 19 | 4228 | ||
| GenBank mRNA | FQ227552.1 | - | 1 | 140 | ||
| FQ227566.1 | - | 1 | 140 | |||
| FQ227617.1 | - | 1 | 140 | |||
| Gene Symbol: LOC683420 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008758873.1 | XP_008757095.1 | 4 | 2722 | ||

Fam120a
Genomic Map