siRNA Details
| Gene Symbol: Tmem258b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001066513.5 | XP_001066513.3 | 1 | 261 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem258b| Gene Symbol: Tmem258b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001066513.5 | XP_001066513.3 | 1 | 261 | ||