siRNA Details
| Gene Symbol: Fam150a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006237796.2 | 2 | 202 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam150a| Gene Symbol: Fam150a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006237796.2 | 2 | 202 | |||