siRNA Details
| Gene Symbol: LOC690097 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008763433.1 | XP_008761655.1 | 2 | 509 | ||
| Gene Symbol: Klra17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001172088.1 | NP_001165559.1 | 1 | 86 | ||

LOC690097
Genomic Map