siRNA Details
| Gene Symbol: Fam47e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008770095.1 | XP_008768317.1 | 6 | 973 | ||
| XM_008770096.1 | XP_008768318.1 | 5 | 880 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam47e| Gene Symbol: Fam47e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008770095.1 | XP_008768317.1 | 6 | 973 | ||
| XM_008770096.1 | XP_008768318.1 | 5 | 880 | |||