siRNA Details
| Gene Symbol: Zdbf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006245044.2 | XP_006245106.1 | 4 | 4898 | ||
| XM_006245045.2 | XP_006245107.1 | 4 | 4881 | |||
| XM_006245046.2 | XP_006245108.1 | 4 | 4928 | |||
| XM_008767157.1 | XP_008765379.1 | 5 | 5200 | |||
| XM_008767158.1 | XP_008765380.1 | 5 | 5221 | |||
| XM_008767159.1 | XP_008765381.1 | 6 | 5283 | |||
| XM_576579.7 | XP_576579.4 | 5 | 5907 | |||

Zdbf2
Genomic Map