siRNA Details
| Gene Symbol: Cpeb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006245978.2 | XP_006246040.1 | 1 | 1143 | ||
| XM_006245979.2 | XP_006246041.1 | 1 | 1143 | |||
| XM_006245980.2 | XP_006246042.1 | 1 | 1143 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cpeb4| Gene Symbol: Cpeb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006245978.2 | XP_006246040.1 | 1 | 1143 | ||
| XM_006245979.2 | XP_006246041.1 | 1 | 1143 | |||
| XM_006245980.2 | XP_006246042.1 | 1 | 1143 | |||