siRNA Details
| Gene Symbol: LOC103689978 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006248475.2 | XP_006248537.1 | 3 | 1751 | ||
| XM_008768779.1 | XP_008767001.1 | 3 | 1752 | |||
| XM_008768780.1 | XP_008767002.1 | 3 | 1748 | |||
| Gene Symbol: RGD1564247 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749241.3 | XP_003749289.1 | 1 | 1736 | ||
| Gene Symbol: Senp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008768807.1 | XP_008767029.1 | 3 | 1752 | ||
| XM_008768808.1 | XP_008767030.1 | 3 | 1748 | |||
| XM_221369.8 | XP_221369.2 | 3 | 1751 | |||

LOC103689978
Genomic Map