siRNA Details
| Gene Symbol: Rab20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109535.1 | NP_001103005.1 | 2 | 1080 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rab20| Gene Symbol: Rab20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109535.1 | NP_001103005.1 | 2 | 1080 | ||