siRNA Details
| Gene Symbol: Lmod3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006236972.2 | XP_006237034.1 | 3 | 1763 | ||
| XM_006236973.2 | XP_006237035.1 | 3 | 1650 | |||
| XM_575617.7 | XP_575617.4 | 2 | 1755 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lmod3| Gene Symbol: Lmod3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006236972.2 | XP_006237034.1 | 3 | 1763 | ||
| XM_006236973.2 | XP_006237035.1 | 3 | 1650 | |||
| XM_575617.7 | XP_575617.4 | 2 | 1755 | |||