siRNA Details
| Gene Symbol: Vps37d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001128193.2 | NP_001121665.1 | 4 | 1220 | ||
| GenBank mRNA | BC166560.1 | - | 4 | 1220 | 7314601 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vps37d| Gene Symbol: Vps37d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001128193.2 | NP_001121665.1 | 4 | 1220 | ||
| GenBank mRNA | BC166560.1 | - | 4 | 1220 | 7314601 | |