siRNA Details
| Gene Symbol: Fam163b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109458.1 | NP_001102928.1 | 3 | 1183 | ||
| XM_006233869.2 | XP_006233931.1 | 2 | 1116 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam163b| Gene Symbol: Fam163b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109458.1 | NP_001102928.1 | 3 | 1183 | ||
| XM_006233869.2 | XP_006233931.1 | 2 | 1116 | |||