siRNA Details
| Gene Symbol: Msrb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001055430.5 | XP_001055430.4 | 7 | 1111 | ||
| XM_006241414.2 | XP_006241476.1 | 8 | 705 | |||
| XM_008765427.1 | XP_008763649.1 | 7 | 1131 | |||
| XM_008765428.1 | XP_008763650.1 | 8 | 755 | |||

Msrb3
Genomic Map